1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine 97%

Supplier: Ambeed
Warning

A140530-10G A140530-1G A140530-250MG A140530-25G A140530-5G
77162-280EA 42.23 USD
77162-280 77162-282 77162-284 77162-286 77162-288
1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine 97%
1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄S
MW: 273.35 g/mol
MDL Number: MFCD27665207
CAS Number: 253168-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR