4-Fluoroveratrole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dimethoxy-4-fluorobenzene, 4-Fluoro-1,2-dimethoxybenzene

A140486-100G A140486-10G A140486-1G A140486-25G A140486-500G A140486-5G
77190-386EA 235.87 USD
77190-386 77190-388 77190-390 77190-392 77190-394 77190-396
4-Fluoroveratrole 98%
4-Fluoroveratrole

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.16 g/mol
MDL Number: MFCD00012201
CAS Number: 398-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR