6-Methoxypicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxypyridine-2-carboxylic acid, 6-Methoxy-2-pyridinecarboxylic acid

A140390-10G A140390-1G A140390-5G A140390-100G A140390-250MG
77162-270EA 59.74 USD
77162-270 77162-272 77566-828 77683-226 77683-227
6-Methoxypicolinic acid 98%
6-Methoxypicolinic acid

Precaución: For research use only.

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD06800962
CAS Number: 26893-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR