1,5-Dimethyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A140353-1G A140353-250MG A140353-25G A140353-5G
77190-374EA 23 USD
77190-374 77190-376 77190-378 77190-380
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid 97%
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD02090876
CAS Number: 31728-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR