1,4-Dibromobutane 99%

Supplier: Ambeed
Danger

Synonyms: 1,4-Dibromo butane

A140270-25G A140270-100G
77190-372EA 6 USD
77190-372 77745-991
1,4-Dibromobutane 99%
1,4-Dibromobutane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₂CH₂Br
MW: 215.92 g/mol
Boiling Pt: 197
Melting Pt: -20
MDL Number: MFCD00000261
CAS Number: 110-52-1
UN: 2922
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR