6-Chloro-2,7-naphthyridin-1(2H)-one 95%

Supplier: Ambeed
Warning

A140194-100MG A140194-250MG A140194-10G A140194-25G A140194-1G A140194-5G
77162-266EA 13 USD
77162-266 77162-268 77566-810 77566-812 77592-662 77891-974
6-Chloro-2,7-naphthyridin-1(2H)-one 95%
6-Chloro-2,7-naphthyridin-1(2H)-one

Precaución: For research use only.

Formula: C₈H₅ClN₂O
MDL Number: MFCD18250873
CAS Number: 1260663-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR