(4-sulfamoylphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A140024-100MG A140024-250MG A140024-5G A140024-1G
77190-356EA 36 USD
77190-356 77190-358 77566-804 77745-990
(4-sulfamoylphenyl)acetic acid 97%
(4-sulfamoylphenyl)acetic acid

Precaución: For research use only.

Formula: C₈H₉NO₄S
MDL Number: MFCD07801100
CAS Number: 22958-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR