2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A139701-100MG A139701-1G A139701-250MG
77162-256EA 63.86 USD
77162-256 77162-258 77162-260
2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanamine hydrochloride 97%
2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀N₄·ClH
MW: 198.653 g/mol
MDL Number: MFCD06739061
CAS Number: 2690-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR