2-(2,2,2-Trifluoroethoxy)pyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A139528-250MG A139528-5G
77190-338EA 170 USD
77190-338 77190-340
2-(2,2,2-Trifluoroethoxy)pyrimidine-5-carboxylic acid 97%
2-(2,2,2-Trifluoroethoxy)pyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₃
MDL Number: MFCD10696903
CAS Number: 944905-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR