4-(3-Fluorophenyl)-2,2-dimethyl-5-(4-(methylthio)phenyl)furan-3(2H)-one 98%

Supplier: Ambeed
Warning

A138958-1G A138958-250MG
77162-244EA 61 USD
77162-244 77162-246
4-(3-Fluorophenyl)-2,2-dimethyl-5-(4-(methylthio)phenyl)furan-3(2H)-one 98%
4-(3-Fluorophenyl)-2,2-dimethyl-5-(4-(methylthio)phenyl)furan-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₉H₁₇FO₂S
MW: 328.40 g/mol
MDL Number: MFCD22570932
CAS Number: 301693-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR