2-(N,N-Dimethylamino)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A138932-1G A138932-250MG A138932-25G A138932-5G
77190-316EA 28.84 USD
77190-316 77190-318 77190-320 77190-322
2-(N,N-Dimethylamino)phenylboronic acid pinacol ester 95%
2-(N,N-Dimethylamino)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂
MW: 247.14 g/mol
MDL Number: MFCD05864304
CAS Number: 832114-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR