Imidazo[1,2-a]pyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A138801-1G A138801-250MG A138801-25G A138801-5G
77162-236EA 29.87 USD
77162-236 77162-238 77162-240 77162-242
Imidazo[1,2-a]pyridine-2-carboxylic acid 98%
Imidazo[1,2-a]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD03419462
CAS Number: 64951-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR