N1,N3-Dibenzylpropane-1,3-diamine 97%

Supplier: Ambeed
Warning

A138307-10G A138307-1G A138307-5G
77190-288EA 53 USD
77190-288 77190-290 77190-292
N1,N3-Dibenzylpropane-1,3-diamine 97%
N1,N3-Dibenzylpropane-1,3-diamine

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂
MW: 254.37 g/mol
MDL Number: MFCD06661418
CAS Number: 10239-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR