6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Thiobis(6-tert-butyl-m-cresol), 4,4'-Thiobis(2-tert-butyl-5-methylphenol), ST029248, Bis(5-tert-butyl-4-hydroxy-2-methylphenyl) sulphide, 4,4'-Thiobis(2-(tert-butyl)-5-methylphenol)

A138125-100G A138125-25G
77190-284EA 20.6 USD
77190-284 77315-574
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol 97%
6,6'-Di-tert-butyl-4,4'-thiodi-m-cresol

Precaución: for Research Use Only

Formula: C₂₂H₃₀O₂S
MW: 358.5374 g/mol
MDL Number: MFCD00026287
CAS Number: 96-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR