Octaphenyloctasilsesquioxane 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A138056-25G A138056-1G A138056-5G
77162-206EA 140 USD
77162-206 77683-212 77891-963
Octaphenyloctasilsesquioxane 98%, Ambeed.Inc
Octaphenyloctasilsesquioxane

Precaución: For research use only.

Formula: C₄₈H₄₀O₁₂Si₈
MDL Number: MFCD00308870
CAS Number: 5256-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR