tert-Butyl (6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A137941-100MG A137941-1G A137941-250MG
77190-274EA 84.46 USD
77190-274 77190-276 77190-278
tert-Butyl (6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl)carbamate 97%
tert-Butyl (6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₁H₂₈BNO₄
MW: 369.26 g/mol
MDL Number: MFCD13191643
CAS Number: 1312611-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR