1,5-Bis(diphenylphosphino)pentane 97%

Supplier: Ambeed
Warning

Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

A137792-100G A137792-10G A137792-1G A137792-25G A137792-5G
77190-254EA 119 USD
77190-254 77190-256 77190-258 77190-260 77190-262
1,5-Bis(diphenylphosphino)pentane 97%
1,5-Bis(diphenylphosphino)pentane

Precaución: for Research Use Only

Formula: C₂₉H₃₀P₂
MDL Number: MFCD00003052
CAS Number: 27721-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR