Methyl 3-methoxy-4-((4-methoxybenzyl)oxy)benzoate 97%
Supplier: Ambeed
|
Warning
|
A137276-100MG
A137276-10G
A137276-250MG
A137276-25G
A137276-5G
77162-166EA
10.3
USD
77162-166
77162-168
77162-170
77162-172
77162-174
Methyl 3-methoxy-4-((4-methoxybenzyl)oxy)benzoate 97%
Methyl 3-methoxy-4-((4-methoxybenzyl)oxy)benzoate
Precaución: for Research Use Only
Formula:
C₁₇H₁₈O₅ MW: 302.32 g/mol |
MDL Number:
MFCD28369484 CAS Number: 1354549-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...