Cerdrulatinib 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Cyclopropylamino)-2-((4-(4-(ethylsulphonyl)piperazin-1-yl)phenyl)amino)pyrimidine-5-carboxamide, Cerdulatinib

A137124-10MG A137124-1MG A137124-50MG A137124-5MG
77162-156EA 50.47 USD
77162-156 77162-158 77162-160 77162-162
Cerdrulatinib 98%
Cerdrulatinib

Precaución: for Research Use Only

Formula: C₂₀H₂₇N₇O₃S
MW: 445.54 g/mol
CAS Number: 1198300-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR