5-Methoxy-2-nitrophenylamine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-methoxy-1-nitrobenzene, 5-Methoxy-2-nitroaniline, 3-Amino-4-nitroanisole

A137083-100G A137083-10G A137083-1G A137083-25G A137083-5G
77190-228EA 200 USD
77190-228 77190-230 77190-232 77190-234 77190-236
5-Methoxy-2-nitrophenylamine 97%
5-Methoxy-2-nitrophenylamine

Precaución: for Research Use Only

Formula: C7H8N2O3
MW: 168.15 g/mol
MDL Number: MFCD00179573
CAS Number: 16133-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR