4,7-Dibromo-1H-pyrrolo[2,3-c]pyridine 96%
Supplier: Ambeed
|
Warning
|
A136997-100MG
A136997-50MG
77190-224EA
114.33
USD
77190-224
77190-226
4,7-Dibromo-1H-pyrrolo[2,3-c]pyridine 96%
4,7-Dibromo-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₇H₄Br₂N₂ MW: 275.93 g/mol |
MDL Number:
MFCD12963396 CAS Number: 619331-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...