NLG919 98%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclohexyl-2-(5H-imidazo[5,1-a]isoindol-5-yl)ethanol, IDO-IN-7

A136625-10MG A136625-1MG A136625-100MG A136625-1G A136625-250MG A136625-5MG
77162-120EA 54.59 USD
77162-120 77162-122 77611-234 77611-236 77611-238 77611-240
NLG919 98%
NLG919

Precaución: for Research Use Only

Formula: C₁₈H₂₂N₂O
MW: 282.38 g/mol
MDL Number: MFCD26142661
CAS Number: 1402836-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR