Phenyl 2-(benzyloxy)-3-(dibenzylamino)-5-fluoro-6-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: TP353

A136576-250MG A136576-1G
77190-218EA 79.31 USD
77190-218 77745-972
Phenyl 2-(benzyloxy)-3-(dibenzylamino)-5-fluoro-6-methylbenzoate 97%
Phenyl 2-(benzyloxy)-3-(dibenzylamino)-5-fluoro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₃₅H₃₀FNO₃
MW: 531.62 g/mol
MDL Number: MFCD28142365
CAS Number: 1253799-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR