1-(trans-4-Pentylcyclohexyl)-4-propylbenzene 97%

Supplier: Ambeed
Warning

A136302-10G A136302-1G A136302-5G
77190-194EA 78.28 USD
77190-194 77190-196 77190-198
1-(trans-4-Pentylcyclohexyl)-4-propylbenzene 97%
1-(trans-4-Pentylcyclohexyl)-4-propylbenzene

Precaución: for Research Use Only

Formula: C₂₀H₃₂
MW: 272.47 g/mol
MDL Number: MFCD11053443
CAS Number: 82991-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR