2-(Chloromethyl)benzonitrile 98%

Supplier: Ambeed
Danger

A136221-100G A136221-500G A136221-25G A136221-10G
77162-098EA 15.45 USD
77162-098 77162-100 77315-174 77683-203
2-(Chloromethyl)benzonitrile 98%
2-(Chloromethyl)benzonitrile

Precaución: For research use only.

Formula: C₈H₆ClN
MW: 151.5929 g/mol
MDL Number: MFCD00019745
CAS Number: 612-13-5
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR