3-Methoxy-2,5-dimethylpyrazine 95%

Supplier: Ambeed
Warning

A136134-1G A136134-250MG
77162-094EA 124.63 USD
77162-094 77162-096
3-Methoxy-2,5-dimethylpyrazine 95%
3-Methoxy-2,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD28162737
CAS Number: 19846-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR