1,2,5-Trifluoro-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A136020-10G A136020-25G
77190-186EA 770 USD
77190-186 77190-188
1,2,5-Trifluoro-3-nitrobenzene 97%
1,2,5-Trifluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₃NO₂
MDL Number: MFCD00456804
CAS Number: 66684-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR