2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%

Supplier: Ambeed
Warning

A135723-100MG A135723-250MG
77190-182EA 57 USD
77190-182 77190-184
2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
MDL Number: MFCD22414580
CAS Number: 1294518-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR