2-Amino-4-nitrobenzaldehyde 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A135608-100MG A135608-5G
77162-070EA 24.72 USD
77162-070 77683-200
2-Amino-4-nitrobenzaldehyde 95%, Ambeed.Inc
2-Amino-4-nitrobenzaldehyde

Precaución: For research use only.

Formula: C₇H₆N₂O₃
MW: 166.13 g/mol
MDL Number: MFCD09264065
CAS Number: 109466-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR