4,4-Dimethoxybutanenitrile 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A135336-1G A135336-25G A135336-5G A135336-100G A135336-500G
77162-064EA 9 USD
77162-064 77162-066 77162-068 77683-195 77891-943
4,4-Dimethoxybutanenitrile 97%, Ambeed.Inc
4,4-Dimethoxybutanenitrile

Precaución: For research use only.

Formula: C₆H₁₁NO₂
MDL Number: MFCD00019883
CAS Number: 14618-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR