Methyl 6-chloro-2-(methylsulfonyl)pyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A134887-100MG A134887-250MG A134887-50MG
77162-048EA 80.34 USD
77162-048 77162-050 77162-052
Methyl 6-chloro-2-(methylsulfonyl)pyrimidine-4-carboxylate 97%
Methyl 6-chloro-2-(methylsulfonyl)pyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₄S
MW: 250.65 g/mol
MDL Number: MFCD16660533
CAS Number: 25742-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR