1,6-Diaminohexane dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1,6-Hexanediamine dihydrochloride, Hexane-1,6-diamine dihydrochloride, Hexamethylenediamine dihydrochloride

A134720-1000G A134720-100G A134720-10G A134720-25G A134720-500G
77190-152EA 59.74 USD
77190-152 77190-154 77190-156 77190-158 77190-160
1,6-Diaminohexane dihydrochloride 97%
1,6-Diaminohexane dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₈Cl₂N₂
MW: 189.13 g/mol
MDL Number: MFCD00012528
CAS Number: 6055-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR