(S)-tert-Butyl 3-(methoxymethyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A134281-100MG A134281-1G A134281-250MG
77162-016EA 106.09 USD
77162-016 77162-018 77683-192
(S)-tert-Butyl 3-(methoxymethyl)piperazine-1-carboxylate 95%
(S)-tert-Butyl 3-(methoxymethyl)piperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₂₂N₂O₃
MW: 230.30 g/mol
MDL Number: MFCD15072107
CAS Number: 955400-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR