(3-Methylbenzofuran-2-yl)boronic acid 97%

Supplier: Ambeed
Warning

A134150-100MG A134150-1G A134150-250MG
77162-014EA 24 USD
77162-014 77549-614 77891-936
(3-Methylbenzofuran-2-yl)boronic acid 97%
(3-Methylbenzofuran-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BO₃
MDL Number: MFCD18383018
CAS Number: 845457-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR