4-((S)-2-((S)-2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)-5-ureidopentanamido)benzyl (4-nitrophenyl) carbonate 98%
Supplier: Ambeed
|
Warning
|
A133919-100MG
A133919-10MG
A133919-1G
A133919-1MG
A133919-250MG
A133919-25MG
A133919-5MG
77161-992EA
44
USD
77161-992
77161-994
77161-996
77161-998
77162-000
77162-002
77162-004
4-((S)-2-((S)-2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)-5-ureidopentanamido)benzyl (4-nitrophenyl) carbonate 98%
4-((S)-2-((S)-2-(6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamido)-3-methylbutanamido)-5-ureidopentanamido)benzyl (4-nitrophenyl) carbonate
Precaución: for Research Use Only
Formula:
C₃₅H₄₃N₇O₁₁ |
MDL Number:
MFCD22200431 CAS Number: 159857-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...