1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)uracil 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-((2R,3S,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil
A133906-100MG
A133906-1G
A133906-250MG
77161-986EA
27.81
USD
77161-986
77161-988
77161-990
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)uracil 95%
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)uracil
Precaución: for Research Use Only
Formula:
C₉H₁₁FN₂O₅ MW: 246.1925 g/mol |
MDL Number:
MFCD00870798 CAS Number: 69123-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...