(3-Methoxypyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A133790-100MG A133790-250MG A133790-1G
77161-972EA 70.04 USD
77161-972 77161-974 77683-188
(3-Methoxypyridin-2-yl)methanol 97%
(3-Methoxypyridin-2-yl)methanol

Precaución: For research use only.

Formula: C₇H₉NO₂
MW: 139.15 g/mol
MDL Number: MFCD08234494
CAS Number: 51984-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR