(11bS)-4-Hydroxy-2,6-di(phenanthren-9-yl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%

Supplier: Ambeed
Warning

A133749-100MG A133749-1G A133749-250MG
77161-966EA 88 USD
77161-966 77161-968 77161-970
(11bS)-4-Hydroxy-2,6-di(phenanthren-9-yl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bS)-4-Hydroxy-2,6-di(phenanthren-9-yl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₄₈H₂₉O₄P
CAS Number: 1043567-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR