1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(1-Ethoxy-ethyl)-4-iodo-1H-pyrazole
A133577-100MG
A133577-250MG
A133577-5G
A133577-1G
77161-960EA
10.3
USD
77161-960
77161-962
77161-964
77683-186
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole 97%
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole
Precaución: For research use only.
Formula:
C₇H₁₁IN₂O MW: 266.08 g/mol |
MDL Number:
MFCD22123601 CAS Number: 575452-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...