1,4-Benzodioxane-6-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Benzodioxane-6-boronic acid, pinacol ester, 2,3-Dihydro-1,4-benzodioxine-6-boronic acid, pinacol ester, 2-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A133428-100MG A133428-250MG A133428-1G
77190-124EA 6 USD
77190-124 77190-126 77745-954
1,4-Benzodioxane-6-boronic acid pinacol ester 98%
1,4-Benzodioxane-6-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MDL Number: MFCD05663890
CAS Number: 517874-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR