(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride
A132601-100MG
A132601-1G
A132601-250MG
77161-942EA
130.81
USD
77161-942
77161-944
77683-177
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 98%
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride
Precaución: For research use only.
Formula:
C₇H₁₄N₂O·2HCl MW: 215.121 g/mol |
MDL Number:
MFCD23105798 CAS Number: 1089280-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...