4-(6-(4-(Piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: LDN-193189 HCl

A132382-10MG A132382-1MG A132382-5MG A132382-50MG A132382-2MG
77161-928EA 109.18 USD
77161-928 77161-930 77161-932 77611-178 77683-172
4-(6-(4-(Piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline hydrochloride 98%
4-(6-(4-(Piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline hydrochloride

Precaución: For research use only.

Formula: C₂₅H₂₃ClN₆
MW: 442.94 g/mol
MDL Number: MFCD17392570
CAS Number: 1062368-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR