2-Methyl-2-(trifluoromethylsulfonamido)propyl methacrylate 95%

Supplier: Ambeed
Warning

A131971-250MG A131971-5G A131971-1G
77161-924EA 13.39 USD
77161-924 77161-926 77683-170
2-Methyl-2-(trifluoromethylsulfonamido)propyl methacrylate 95%
2-Methyl-2-(trifluoromethylsulfonamido)propyl methacrylate

Precaución: For research use only.

Formula: C₉H₁₄F₃NO₄S
MW: 289.27 g/mol
MDL Number: MFCD28369489
CAS Number: 1268257-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR