1-(3-Fluorophenyl)cyclopropanamine 98%

Supplier: Ambeed
Warning

A131867-100MG A131867-1G A131867-250MG
77161-912EA 39.14 USD
77161-912 77161-914 77161-916
1-(3-Fluorophenyl)cyclopropanamine 98%
1-(3-Fluorophenyl)cyclopropanamine

Precaución: for Research Use Only

Formula: C₉H₁₀FN
MW: 151.18 g/mol
MDL Number: MFCD07374478
CAS Number: 764647-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR