4-Isopropoxy-3-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A131759-100MG A131759-1G A131759-250MG A131759-5G
77161-904EA 38.11 USD
77161-904 77161-906 77161-908 77161-910
4-Isopropoxy-3-(trifluoromethyl)benzoic acid 97%
4-Isopropoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
MW: 248.20 g/mol
MDL Number: MFCD12913405
CAS Number: 213598-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR