cis-1-Boc-3-fluoro-4-hydroxypiperidine 95%

Supplier: Ambeed
Warning

Synonyms: (3.4)-cis-3-Fluoro-4-hydroxy-piperidine-1-carboxylic acid tert-Butyl ester, cis-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

A131433-1G A131433-250MG A131433-25G A131433-5G
77161-896EA 45.32 USD
77161-896 77161-898 77161-900 77161-902
cis-1-Boc-3-fluoro-4-hydroxypiperidine 95%
cis-1-Boc-3-fluoro-4-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₀H₁₈FNO₃
MW: 219.26 g/mol
MDL Number: MFCD18791209
CAS Number: 955028-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR