1-(4-Fluorophenyl)piperazin-2-one hydrochloride 95%

Supplier: Ambeed
Warning

A131369-100MG A131369-1G A131369-250MG
77161-890EA 36.05 USD
77161-890 77161-892 77161-894
1-(4-Fluorophenyl)piperazin-2-one hydrochloride 95%
1-(4-Fluorophenyl)piperazin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFN₂O
MW: 230.66 g/mol
MDL Number: MFCD27922068
CAS Number: 697305-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR