4-Cyano-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A130431-1G A130431-250MG A130431-25G A130431-5G
77161-856EA 10 USD
77161-856 77161-858 77161-860 77891-916
4-Cyano-3-nitrobenzoic acid 98%
4-Cyano-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄N₂O₄
MDL Number: MFCD16876894
CAS Number: 153775-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR