3-Acetyl-2-fluoropyridine 97%

Supplier: Ambeed
Warning

A130329-100MG A130329-1G A130329-250MG A130329-5G
77161-850EA 10 USD
77161-850 77161-852 77161-854 77891-915
3-Acetyl-2-fluoropyridine 97%
3-Acetyl-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD07375072
CAS Number: 79574-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR