(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-hydroxyphenethoxy)tetrahydro-2H-pyran-3,4,5-triol 98%

Supplier: Ambeed
Warning

A130275-10MG A130275-1G A130275-25G A130275-5G A130275-5MG
77190-030EA 6 USD
77190-030 77190-032 77190-034 77190-036 77190-038
(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-hydroxyphenethoxy)tetrahydro-2H-pyran-3,4,5-triol 98%
(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-hydroxyphenethoxy)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₇
MDL Number: MFCD00210553
CAS Number: 10338-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR